Merck
  • Target-selective drug delivery through liposomes labeled with oligobranched neurotensin peptides.
Target-selective drug delivery through liposomes labeled with oligobranched neurotensin peptides.
ChemMedChem (2011-03-04)
Chiara Falciani, Antonella Accardo, Jlenia Brunetti, Diego Tesauro, Barbara Lelli, Alessandro Pini, Luisa Bracci, Giancarlo Morelli
ABSTRACT
The structure and the in vitro behavior of liposomes filled with the cytotoxic drug doxorubicin (Doxo) and functionalized on the external surface with a branched moiety containing four copies of the 8-13 neurotensin (NT) peptide is reported. The new functionalized liposomes, DOPC-NT₄Lys(C₁₈)₂, are obtained by co-aggregation of the DOPC phospholipid with a new synthetic amphiphilic molecule, NT₄ Lys(C₁₈)₂, which contains a lysine scaffold derivatized with a lipophilic moiety and a tetrabranched hydrophilic peptide, NT8-13, a neurotensin peptide fragment well known for its ability to mimic the neurotensin peptide in receptor binding ability. Dynamic light scattering measurements indicate a value for the hydrodynamic radius (RH) of 88.3±4.4 nm. The selective internalization and cytotoxicity of DOPC-NT₄ Lys(C₁₈)₂ liposomes containing Doxo, as compared to pure DOPC liposomes, were tested in HT29 human colon adenocarcinoma and TE671 human rhabdomyosarcoma cells, both of which express neurotensin receptors. Peptide-functionalized liposomes show a clear advantage in comparison to pure DOPC liposomes with regard to drug internalization in both HT29 and TE671 tumor cells: FACS analysis indicates an increase in fluorescence signal of the NT₄-liposomes, compared to the DOPC pure analogues, in both cell lines; cytotoxicity of DOPC-NT₄ Lys(C₁₈)₂-Doxo liposomes is increased four-fold with respect to DOPC-Doxo liposomes in both HT29 and TE671 cell lines. These effects could to be ascribed to the higher rate of internalization for DOPC-NT₄ Lys(C₁₈)₂-Doxo liposomes, due to stronger binding driven by a lower dissociation constant of the NT₄-liposomes that bind the membrane onto a specific protein, in contrast to DOPC liposomes, which approach the plasma membrane unselectively.
Sigma-Aldrich
Neurotensin Fragment 8-13 acetate salt, ≥97% (HPLC)