Merck
  • Properties of mouse vomeronasal receptor and assessment of its role in pheromone signalling.
Properties of mouse vomeronasal receptor and assessment of its role in pheromone signalling.
Rapid communications in mass spectrometry : RCM (2010-12-31)
Soundarapandian Kannan, Pandi Mareeswaran, Muthukalingan Krishnan, Shanmugam Achiraman, Neil Spencer, Balakrishnan F Nytingel
ABSTRACT
Vomeronasal type 2 receptor (V2Rx) from Swiss mouse (Mus musculus (L.)) was analyzed by high-resolution ion-exchange chromatography, reversed-phase high-performance liquid chromatography (RP-HPLC), matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF-MS), Fourier transform ion cyclotron resonance mass spectrometry (FTICR-MS), Ion Spray tandem mass spectrometry (MS/MS), 10% sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS-PAGE) and 1-aminoanthracene (1-AMA) fluorometric assay. Vomeronasal sensory neuronal cell bound proteins were resolved into major protein peaks. Several proteins were identified and subsequently purified as the V2Rx receptor on 10% SDS-PAGE with trace amounts of other protein bands. The molecular weight of the identified V2Rx was 109 kDa. MALDI-TOF and micro-sequencing experiments demonstrated that the identified V2Rx receptor shared considerable sequence similarity with vomeronasal receptor type 2 (NCBI Accession Number AB267725), which is a seven transmembrane peptide with 912 amino acid residues. The molecular characterization revealed that the N-terminus of the V2Rx receptor contained the 11GAEAAE16 domain involved in pheromone signalling. The biometric assay (octanamine-V2Rx binding) showed the identified V2Rx receptor and mouse sex pheromone to 2-octanamine (methyl heptyl) in a 1:1 ratio. Uptake of odourants determined in physiological condition showed enhanced V2Rx receptors as volatile hydrophobic pheromone receptors in the vomeronasal neuron of the Swiss mouse.
Sigma-Aldrich
1-Aminoanthracene, technical grade, 90%