Merck
  • Isolation of N,N-dimethyl and N-methylserotonin 5-O-β-glucosides from immature Zanthoxylum piperitum seeds.
Isolation of N,N-dimethyl and N-methylserotonin 5-O-β-glucosides from immature Zanthoxylum piperitum seeds.
Bioscience, biotechnology, and biochemistry (2010-09-14)
Emiko Yanase, Masaki Ohno, Hironari Harakawa, Shin-ichi Nakatsuka
ABSTRACT
Two serotonin derivatives, N,N-dimethylserotonin 5-O-β-glucoside (1a) and N-methylserotonin 5-O-β-glucoside (1b) were isolated from immature seeds of Zanthoxylum piperitum. Their structures were determined by multi-step conversion reactions and spectroscopic analyses. Immature seeds of Z. piperitum contained extremely high levels of compounds 1a and 1b of approximately 0.29% and 0.15% (w/w), respectively.
Sigma-Aldrich
5-Hydroxy--methyltryptamine oxalate, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nω-Methyl-5-hydroxy­trypt­amine oxalate salt, ≥90% (HPLC)