Merck
  • [Determination of volatile organic compounds in lung cancer cell lines and lung cancer tissue].
[Determination of volatile organic compounds in lung cancer cell lines and lung cancer tissue].
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences (2010-06-15)
Yan-jie Hu, Yuan-hua Qiu, En-guo Chen, Ke-jing Ying, Jin Yu, Ping Wang
ABSTRACT
To identify the volatile organic compounds (VOCs) in lung cancer tissue and lung cancer cell lines. The lung cancer tissue samples from 18 patients were cultured and 4 lung cell lines (A549, NCI-H446, SK-MES-1, BEAS-2B) were also included in the study. Air samples in the headspace of culture flasks were analyzed for VOCs with solid-phase micro-extraction and gas chromatography-mass spectroscopy technique (SPME-GC/MS). Two kinds of VOCs 2-pentadecanone and nonadecane were detected in lung cancer cell lines A549, NCI-H446 and SK-MES-1. The concentration of 2-pentadecanone were (1.382 + or -0.171) X 10(-5)mg/L, (1.681 + or - 0.190) X 10(-4)mg/L and (2.835 + or - 0.401) X 10(-6)mg/L, respectively; the concentrations of nonadecane were (8.382 + or - 0.606 ) X 10(-6)mg/L, (1.845 + or - 0.130) X 10(-5)mg/L and (6.220 + or - 0.362) X 10(-6)mg/L), respectively. The eicosane was detected in A549 and NCI-H446 with the concentration of (8.313 + or - 1.130) X 10(-6)mg/L and (1.020 + or - 0.141) X 10(-5)mg/L), respectively. All the 3 VOCs were not detected in cell line BEAS-2B. The concentrations of 12 VOCs including decane, 2- pentadecanone, nonadecane and eicosane were high in 18 lung cancer tissue samples; the concentrations of 2-pentadecanone were 5.421 X 10(-6)mg/L-3.621 X 10(-5)mg/L,those of nonadecane were 5.805 X 10(-6)mg/L-1.830 X 10(-5)mg/L, those of eicosane were 2.730 X 10(-6)mg/L-2.343 X 10(-5)mg/L. There were no differences of VOCs levels among patients with different cancer differentiation (P>0.05). The concentration of eicosane in the non-squamous carcinoma was higher than that in squamous carcinoma, the same results were confirmed in the lung cancer cell lines. This study has identified VOCs produced by lung cancer tissue, which may support to use breath test as a complementary noninvasive diagnostic method for lung cancer.
Sigma-Aldrich
Nonadecane, 99%
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #5, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
Nonadecane, analytical standard
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, white hub
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Sampling Stand, for use with 15 mL vials