Merck
  • Bioreduction of alpha-methylcinnamaldehyde derivatives: chemo-enzymatic asymmetric synthesis of Lilial and Helional.
Bioreduction of alpha-methylcinnamaldehyde derivatives: chemo-enzymatic asymmetric synthesis of Lilial and Helional.
Dalton transactions (Cambridge, England : 2003) (2010-05-13)
Clemens Stueckler, Nicole J Mueller, Christoph K Winkler, Silvia M Glueck, Karl Gruber, Georg Steinkellner, Kurt Faber
ABSTRACT
Nonracemic aryl-substituted alpha-methyldihydrocinnamaldehyde derivatives employed as olfactory principles in perfumes (Lilial, Helional) were obtained via enzymatic reduction of the corresponding cinnamaldehyde precursors using cloned and overexpressed ene-reductases. (R)-Enantiomers were obtained using the old-yellow-enzyme (OYE) homolog YqjM from Bacillus subtilis and 12-oxophytodienoic acid reductase isoenzyme OPR1 from tomato (e.e.(max) 53%), and (S)-aldehydes were furnished in up to 97% e.e. using isoenzyme OPR3, nicotinamide 2-cyclohexene-1-one reductase NCR from Zymomonas mobilis and yeast OYE isoenzymes 1-3 under optimised reaction conditions in the presence of t-butyl methyl ether as the co-solvent. The stereochemical outcome of the reduction of alpha-methylcinnamaldehyde using NCR and OYEs 1-3 [previously reported to be (R)] was unambiguously corrected to be (S).