Merck
  • Dissolved organic matter enhances transport of PAHs to aquatic organisms.
Dissolved organic matter enhances transport of PAHs to aquatic organisms.
Environmental science & technology (2009-10-24)
Thomas L Ter Laak, Martin A Ter Bekke, Joop L M Hermens
ABSTRACT
In this study, the uptake of pyrene and benzo[b]fluoranthene by an aquatic worm (Lumbriculus variegatus) and a poly(dimethylsiloxane) coated glass fiber was studied at different humic acid concentrations. The accumulation of pyrene was not affected by the presence of the humic matrix. However, the accumulation rate of benzo[b]fluoranthene increased a factor of 3 for the fiber and a factor of 4 when 55 mg L(-1) dissolved organic carbon was added in the form of humic acid. The difference between the two chemicals can be explained by the higher affinity of benzo[b]fluoranthene for the dissolved humic material. A comparison of modeled transport enhancement of benzo[b]fluoranthene by humic acid and the experimental results suggested that the benzo[b]fluoranthene complexed with the humic phase was not completely labile.
Sigma-Aldrich
Benzo[b]fluoranthene, 98%
Supelco
Benzo[b]fluoranthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzo[b]fluoranthene, analytical standard