Merck
Quantitative plasma biomarker analysis in HDI exposure assessment.
The Annals of occupational hygiene (2009-10-07)
Sheila L Flack, Kenneth W Fent, Linda G Trelles Gaines, Jennifer M Thomasen, Steve Whittaker, Louise M Ball, Leena A Nylander-French
ABSTRACT
Quantification of amines in biological samples is important for evaluating occupational exposure to diisocyanates. In this study, we describe the quantification of 1,6-hexamethylene diamine (HDA) levels in hydrolyzed plasma of 46 spray painters applying 1,6-hexamethylene diisocyanate (HDI)-containing paint in vehicle repair shops collected during repeated visits to their workplace and their relationship with dermal and inhalation exposure to HDI monomer. HDA was detected in 76% of plasma samples, as heptafluorobutyryl derivatives, and the range of HDA concentrations was < or =0.02-0.92 microg l(-1). After log-transformation of the data, the correlation between plasma HDA levels and HDI inhalation exposure measured on the same workday was low (N = 108, r = 0.22, P = 0.026) compared with the correlation between plasma HDA levels and inhalation exposure occurring approximately 20 to 60 days before blood collection (N = 29, r = 0.57, P = 0.0014). The correlation between plasma HDA levels and HDI dermal exposure measured on the same workday, although statistically significant, was low (N = 108, r = 0.22, P = 0.040) while the correlation between HDA and dermal exposure occurring approximately 20 to 60 days before blood collection was slightly improved (N = 29, r = 0.36, P = 0.053). We evaluated various workplace factors and controls (i.e. location, personal protective equipment use and paint booth type) as modifiers of plasma HDA levels. Workers using a downdraft-ventilated booth had significantly lower plasma HDA levels relative to semi-downdraft and crossdraft booth types (P = 0.0108); this trend was comparable to HDI inhalation and dermal exposure levels stratified by booth type. These findings indicate that HDA concentration in hydrolyzed plasma may be used as a biomarker of cumulative inhalation and dermal exposure to HDI and for investigating the effectiveness of exposure controls in the workplace.
Sigma-Aldrich
Hexamethylenediamine, technical grade, 70%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexamethylenediamine, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexamethylenediamine dihydrochloride, 99%