Merck
  • Modelling the removal of p-TSA (para-toluenesulfonamide) during rapid sand filtration used for drinking water treatment.
Modelling the removal of p-TSA (para-toluenesulfonamide) during rapid sand filtration used for drinking water treatment.
Water research (2009-09-22)
Raffaella Meffe, Claus Kohfahl, Ekkehard Holzbecher, Gudrun Massmann, Doreen Richter, Uwe Dünnbier, Asaf Pekdeger
ABSTRACT
A finite element model was set-up to determine degradation rate constants for p-TSA during rapid sand filtration (RSF). Data used for the model originated from a column experiment carried out in the filter hall of a drinking water treatment plant in Berlin (Germany). Aerated abstracted groundwater was passed through a 1.6m long column-shaped experimental sand filter applying infiltration rates from 2 to 6mh(-1). Model results were fitted to measured profiles and breakthrough curves of p-TSA for different infiltration rates using both first-order reaction kinetics and Michaelis-Menten kinetics. Both approaches showed that degradation rates varied both in space and time. Higher degradation rates were observed in the upper part of the column, probably related to higher microbial activity in this zone. Measured and simulated breakthrough curves revealed an adaption phase with lower degradation rates after infiltration rates were changed, followed by an adapted phase with more elevated degradation rates. Irrespective of the mathematical approach and the infiltration rate, degradation rates were very high, probably owing to the fact that filter sands have been in operation for decades, receiving high p-TSA concentrations with the raw water.
Sigma-Aldrich
p-Toluenesulfonamide, reagent grade, 97%
Sigma-Aldrich
p-Toluenesulfonamide, ReagentPlus®, ≥99%