Merck
  • Pharmacokinetics of temozolomide administered in combination with O6-benzylguanine in children and adolescents with refractory solid tumors.
Pharmacokinetics of temozolomide administered in combination with O6-benzylguanine in children and adolescents with refractory solid tumors.
Cancer chemotherapy and pharmacology (2009-05-12)
Holly J Meany, Katherine E Warren, Elizabeth Fox, Diane E Cole, Alberta A Aikin, Frank M Balis
ABSTRACT
Temozolomide pharmacokinetics were evaluated in children receiving concurrent O(6)-benzylguanine (O(6)BG), which enhanced the hematological toxicity of temozolomide. Temozolomide was administered orally, daily for 5 days starting at 28 mg/m(2) per day with escalations to 40, 55, 75 and 100 mg/m(2) per day with O(6)BG intravenously daily for 5 days at doses of 60, 90 or 120 mg/m(2) per day. Plasma samples were drawn over 48 h after the day 5 dose. Temozolomide was quantified with a validated HPLC/tandem mass spectroscopic assay. Temozolomide was rapidly absorbed (mean T (max), 2.1 h). The mean apparent clearance (CL/F) (96 mL/min/m(2)) was similar to the CL/F for temozolomide alone and was not age- or gender-dependent. There was minimal inter-patient variability. The enhanced hematologic toxicity resulting from combining O(6)BG with temozolomide does not appear to be the result of a pharmacokinetic interaction between the agents.
Sigma-Aldrich
O6-Benzylguanine, ≥98% (TLC), solid