Merck
  • Photodynamic therapy with aluminum-chloro-phthalocyanine induces necrosis and vascular damage in mice tongue tumors.
Photodynamic therapy with aluminum-chloro-phthalocyanine induces necrosis and vascular damage in mice tongue tumors.
Journal of photochemistry and photobiology. B, Biology (2008-12-23)
João Paulo Figueiró Longo, Silene Paulino Lozzi, Andreza Ribeiro Simioni, Paulo César Morais, Antônio Cláudio Tedesco, Ricardo Bentes Azevedo
ABSTRACT
In this paper we describe the efficacy of the liposomal-AlClPc (aluminum-chloro-phthalocyanine) formulation in PDT study against Ehrlich tumor cells proliferation in immunocompetent swiss mice tongue. Experiments were conduced in sixteen tumor induced mice that were divided in three control groups: (1) tumor without treatment; (2) tumor with 100J/cm(2) laser (670nm) irradiation; and (3) tumor with AlClPc peritumoral injection; and a PDT experimental group when tumors received AlClPc injection followed by tumor irradiation. Control groups present similar macroscopically and histological patterns after treatments, while PDT treatment induced 90% of Ehrlich tumor necrosis after 24h of one single application, showing the efficacy of liposome-AlClPc (aluminum-chloro-phthalocyanine) mediated PDT on the treatment of oral cancer.
Sigma-Aldrich
Aluminum phthalocyanine chloride, Dye content ~85 %