Merck
  • Comparison of the novel subtype-selective GABAA receptor-positive allosteric modulator NS11394 [3'-[5-(1-hydroxy-1-methyl-ethyl)-benzoimidazol-1-yl]-biphenyl-2-carbonitrile] with diazepam, zolpidem, bretazenil, and gaboxadol in rat models of inflammatory and neuropathic pain.
Comparison of the novel subtype-selective GABAA receptor-positive allosteric modulator NS11394 [3'-[5-(1-hydroxy-1-methyl-ethyl)-benzoimidazol-1-yl]-biphenyl-2-carbonitrile] with diazepam, zolpidem, bretazenil, and gaboxadol in rat models of inflammatory and neuropathic pain.
The Journal of pharmacology and experimental therapeutics (2008-09-16)
G Munro, J A Lopez-Garcia, I Rivera-Arconada, H K Erichsen, E Ø Nielsen, J S Larsen, P K Ahring, N R Mirza
ABSTRACT
Spinal administration of GABA(A) receptor modulators, such as the benzodiazepine drug diazepam, partially alleviates neuropathic hypersensitivity that manifests as spontaneous pain, allodynia, and hyperalgesia. However, benzodiazepines are hindered by sedative impairments and other side effect issues occurring mainly as a consequence of binding to GABA(A) receptors containing the alpha(1) subunit. Here, we report on the novel subtype-selective GABA(A) receptor-positive modulator NS11394 [3'-[5-(1-hydroxy-1-methyl-ethyl)-benzoimidazol-1-yl]-biphenyl-2-carbonitrile], which possesses a functional efficacy selectivity profile of alpha(5) > alpha(3) > alpha(2) > alpha(1) at GABA(A) alpha subunit-containing receptors. Oral administration of NS11394 (1-30 mg/kg) to rats attenuated spontaneous nociceptive behaviors in response to hindpaw injection of formalin and capsaicin, effects that were blocked by the benzodiazepine site antagonist flumazenil. Ongoing inflammatory nociception, observed as hindpaw weight-bearing deficits after Freund's adjuvant injection, was also completely reversed by NS11394. Likewise, hindpaw mechanical allodynia was fully reversed by NS11394 in two rat models of peripheral neuropathic pain. Importantly, NS11394-mediated antinociception occurred at doses 20 to 40-fold lower than those inducing minor sedative or ataxic impairments. In contrast, putative antinociception associated with administration of either diazepam, zolpidem, or gaboxadol only occurred at doses producing intolerable side effects, whereas bretazenil was completely inactive despite minor influences on motoric function. In electrophysiological studies, NS11394 selectively attenuated spinal nociceptive reflexes and C-fiber-mediated wind-up in vitro pointing to involvement of a spinal site of action. The robust therapeutic window seen with NS11394 in animals suggests that compounds with this in vitro selectivity profile could have potential benefit in clinical treatment of pain in humans.
Sigma-Aldrich
Bretazenil, ≥96% (HPLC), solid