Merck
  • The dihydrolipoamide dehydrogenase of Aeromonas caviae ST exhibits NADH-dependent tellurite reductase activity.
The dihydrolipoamide dehydrogenase of Aeromonas caviae ST exhibits NADH-dependent tellurite reductase activity.
Biochemical and biophysical research communications (2008-08-05)
Miguel E Castro, Roberto Molina, Waldo Díaz, Sergio E Pichuantes, Claudio C Vásquez
ABSTRACT
Potassium tellurite (K(2)TeO(3)) is extremely toxic for most forms of life and only a limited number of organisms are naturally resistant to the toxic effects of this compound. Crude extracts prepared from the environmental isolate Aeromonas caviae ST catalize the in vitro reduction of TeO32- in a NADH-dependent reaction. Upon fractionation by ionic exchange column chromatography three major polypeptides identified as the E1, E2, and E3 components of the pyruvate dehydrogenase (PDH) complex were identified in fractions exhibiting tellurite-reducing activity. Tellurite reductase and pyruvate dehydrogenase activities co-eluted from a Sephadex gel filtration column. To determine which component(s) of the PDH complex has tellurite reductase activity, the A. caviae ST structural genes encoding for E1 (aceE), E2 (aceF), and E3 (lpdA) were independently cloned and expressed in Escherichia coli and their gene products purified. Results indicated that tellurite reductase activity lies almost exclusively in the E3 component, dihydrolipoamide dehydrogenase. The E3 component of the PDH complex from E. coli, Zymomonas mobilis, Streptococcus pneumoniae, and Geobacillus stearothermophilus also showed NADH-dependent tellurite reductase in vitro suggesting that this enzymatic activity is widely distributed among microorganisms.
Product Description
Millipore
Potassium tellurite solution, 1% in H2O, for Staphylococcus spp., for Corynebacterium spp., tube of 10 × 10 mL