Merck
  • Binding of orthosteric ligands to the allosteric site of the M(2) muscarinic cholinergic receptor.
Binding of orthosteric ligands to the allosteric site of the M(2) muscarinic cholinergic receptor.
Molecular pharmacology (2008-06-17)
Dar'ya S Redka, Luca F Pisterzi, James W Wells
ABSTRACT
The M(2) muscarinic receptor has two topographically distinct sites: the orthosteric site and an allosteric site recognized by compounds such as gallamine. It also can exhibit cooperative effects in the binding of orthosteric ligands, presumably to the orthosteric sites within an oligomer. Such effects would be difficult to interpret, however, if those ligands also bound to the allosteric site. Monomers of the hemagglutinin (HA)- and FLAG-tagged human M(2) receptor therefore have been purified from coinfected Sf9 cells and examined for any effect of the antagonist N-methyl scopolamine or the agonist oxotremorine-M on the rate at which N-[(3)H]methyl scopolamine dissociates from the orthosteric site (k(obsd)). The predominantly monomeric status was confirmed by coimmunoprecipitation and by cross-linking with bis(sulfosuccinimidyl)suberate. Both N-methyl scopolamine and oxotremorine-M acted in a cooperative manner to decrease k(obsd) by 4.5- and 9.1-fold, respectively; the corresponding estimates of affinity (log K(L)) are -2.55 +/- 0.13 and -2.29 +/- 0.14. Gallamine and the allosteric ligand obidoxime decreased k(obsd) by more than 100-fold (log K(L) = -4.12 +/- 0.04) and by only 1.1-fold (log K(L) = -1.73 +/- 0.91), respectively. Obidoxime reversed the effect of N-methyl scopolamine, oxotremorine-M, and gallamine in a manner that could be described by a model in which all four ligands compete for a common allosteric site. Ligands generally assumed to be exclusively orthosteric therefore can act at the allosteric site of the M(2) receptor, albeit at comparatively high concentrations.
Sigma-Aldrich
Gallamine triethiodide, ≥98% (TLC), powder, muscarinic receptor antagonist