Merck
  • Rapid determination of carbamate pesticides in food using dual counter-current chromatography directly interfaced with mass spectrometry.
Rapid determination of carbamate pesticides in food using dual counter-current chromatography directly interfaced with mass spectrometry.
Journal of chromatography. A (2008-02-26)
Yuko Ito, Tomomi Goto, Sadaji Yamada, Tsutomu Ohno, Hiroshi Matsumoto, Hisao Oka, Yoichiro Ito
ABSTRACT
Dual counter-current chromatography (dual CCC)-tandem mass spectrometry (MS/MS) was successfully performed with a newly designed spiral column for dual CCC. The small column capacity required for directly coupling with electrospray MS/MS was accomplished by forming a rectangular spiral groove on a plastic disk and sealing it with a PTFE sheet. This novel dual CCC-MS/MS technique was successfully applied for the rapid determination of methomyl, fenobucarb and carbaryl pesticides in food. A two-phase solvent system of n-hexane-acetonitrile-0.1% formic acid (45:45:10) was suitable for both good dual CCC separation and sufficient ionization of pesticides. Recoveries of these three pesticides from mandarin orange and spinach samples fortified at 0.05 mg/kg were in the range of 93-107% with relative standard deviations of 2.4-3.8%.