Merck
[Lavomax in outpatient treatment of urogenital chlamidiasis in males].
Urologiia (Moscow, Russia : 1999) (2008-02-08)
I G Leont'ev
ABSTRACT
Sixty four males (age 21-45 years) with urogenital chlamidiasis were divided into two groups. 34 patients of the study group received interferon inductor and lavomax. 30 patients of the control group--interferon inductor and cycloferon. Treatment efficacy in the study group was higher.
Sigma-Aldrich
Tilorone dihydrochloride, 95%