Merck
  • Comparative effects of different triazole compounds on growth, photosynthetic pigments and carbohydrate metabolism of Solenostemon rotundifolius.
Comparative effects of different triazole compounds on growth, photosynthetic pigments and carbohydrate metabolism of Solenostemon rotundifolius.
Colloids and surfaces. B, Biointerfaces (2007-08-03)
A Kishorekumar, C Abdul Jaleel, P Manivannan, B Sankar, R Sridharan, R Panneerselvam
ABSTRACT
The effects of two triazole compounds, triadimefon and hexaconazole, on the growth and carbohydrate metabolism were studied in Solenostemon rotundifolius Poir., Morton plants under pot culture. Plants were treated with triadimefon at 15mg l(-1) and hexaconazole at 10mg l(-1) separately by soil drenching on 80, 110 and 140 days after planting (DAP). The plants were harvested randomly and growth parameters were studied on 90, 120 and 150 DAP for determining the effect of both the triazole on growth and chlorophyll pigments. These triazole compounds increased the chlorophyll pigments. However, both the treatments decreased the fresh and dry weights of shoot and leaf area. Both these triazole resulted in a marginal increase in starch content and decreased the sugar contents. The carbohydrate metabolizing enzymes alpha- and beta-amylase activities were reduced and invertase activity increased in S. rotundifolius under triadimefon and hexaconazole treatments.