Merck
  • Kinetic manifestation of processivity during multiple methylations catalyzed by SET domain protein methyltransferases.
Kinetic manifestation of processivity during multiple methylations catalyzed by SET domain protein methyltransferases.
Biochemistry (2007-03-07)
Lynnette M A Dirk, E Megan Flynn, Kevin Dietzel, Jean-François Couture, Raymond C Trievel, Robert L Houtz
ABSTRACT
Processive versus distributive methyl group transfer was assessed for pea Rubisco large subunit methyltransferase, a SET domain protein lysine methyltransferase catalyzing the formation of trimethyllysine-14 in the large subunit of Rubisco. Catalytically competent complexes between an immobilized form of des(methyl) Rubisco and Rubisco large subunit methyltransferase were used to demonstrate enzyme release that was co-incident with and dependent on formation of trimethyllysine. Catalytic rate constants determined for formation of trimethyllysine were considerably lower ( approximately 10-fold) than rate constants determined for total radiolabel incorporation from [3H-methyl]-S-adenosylmethionine. Double-reciprocal velocity plots under catalytic conditions favoring monomethyllysine indicated a random or ordered reaction mechanism, while conditions favoring trimethyllysine suggested a hybrid ping-pong mechanism. These results were compared with double-reciprocal velocity plots and product analyses obtained for HsSET7/9 (a monomethyltransferase) and SpCLR4 (a dimethyltransferase) and suggest a predictive ability of double-reciprocal velocity plots for single versus multiple methyl group transfers by SET domain protein lysine methyltransferases. A model is proposed for SET domain protein lysine methyltransferases in which initial binding of polypeptide substrate and S-adenosylmethionine is random, with polypeptide binding followed by deprotonation of the epsilon-amine of the target lysyl residue and subsequent methylation. Following methyl group transfer, S-adenosylhomocysteine and monomethylated polypeptide dissociate from monomethyltransferases, but di- and trimethyltransferases begin a successive and catalytically obligatory deprotonation of enzyme-bound methylated lysyl intermediates, which along with binding and release of S-adenosylmethionine and S-adenosylhomocysteine is manifested as a hybrid ping-pong-like reaction mechanism.
Sigma-Aldrich
Nε,Nε,Nε-Trimethyllysine hydrochloride, ≥97% (TLC)