Merck
  • Frequency upconversion in Pr3+-Li2O-TeO2 binary glass by decay curve analysis.
Frequency upconversion in Pr3+-Li2O-TeO2 binary glass by decay curve analysis.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-03-01)
Vineet Kumar Rai, S B Rai
ABSTRACT
Using the decay curve analysis frequency upconversion under continuous wave NIR laser radiation ( approximately 890 nm) following the non-resonant excitation at higher energies than 1G4 level has been reported. The decay curve of the antistokes emission (3P0-->3H4) shows similar behavior as that of the stokes emission (3P0-->3H4) and does not show any rise time attributing to the excited state antistokes absorption assisted by phonon emission. The covalency, bonding parameters and nephalauxetic effect for the present system has also been determined.
Sigma-Aldrich
Tellurium dioxide, ≥99%
Sigma-Aldrich
Tellurium dioxide, 99.995% trace metals basis
Sigma-Aldrich
Tellurium dioxide, ≥97.0%