Merck
  • Developmental toxic effects of antifungal flusilazole administered by gavage to mice.
Developmental toxic effects of antifungal flusilazole administered by gavage to mice.
Birth defects research. Part B, Developmental and reproductive toxicology (2006-12-26)
Amina T Farag, Hala H Ibrahim
ABSTRACT
The developmental toxicity of flusilazole was studied in CD-1 mice after oral administration. Pregnant mice were given flusilazole at doses of 0 (corn oil), 10, 20, and 40 mg/kg/day, by gavage, on gestational days (GD) 6-15. Maternal toxicity, as evidenced by reduction in body weight gain and signs of toxicity, was observed at the middle- and high-dose groups. No significant incidence of resorptions or death was observed in any of dose groups. There was a pronounced reduction in fetal weight, which was significantly lower than control from 20 and 40 mg/kg/day. There was no significant increase in the incidence of fetuses with external or visceral malformations in any of dose groups, but there was a significant increase in the incidence of skeletal malformations was observed at 20 and 40 mg/kg/day. The results of this study reported marked maternal toxicity, growth retardation, and skeletal abnormalities in the mid- and high-dose groups. It seems likely that marked maternal toxicity contributed to the observed alterations in fetal growth retardation and skeletal development. The no-observed-effect level in the present study for maternal and developmental toxicity was 10 mg/kg/day.