Merck
  • Cyclooxygenase isozymes are expressed in human myeloma cells but not involved in anti-proliferative effect of cyclooxygenase inhibitors.
Cyclooxygenase isozymes are expressed in human myeloma cells but not involved in anti-proliferative effect of cyclooxygenase inhibitors.
Molecular carcinogenesis (2005-12-31)
Jie Ding, Kazuhito Tsuboi, Hiroshi Hoshikawa, Rieko Goto, Nozomu Mori, Michiko Katsukawa, Emi Hiraki, Shozo Yamamoto, Masahiro Abe, Natsuo Ueda
ABSTRACT
Considering possible tumorigenic activity of cyclooxygenase (COX) isozymes in myeloma, we examined expression levels of COX-1 and -2 in seven human myeloma cell lines (ARH-77, IM-9, RPMI-8226, HPC, HS-Sultan, TSPC-1, and U-266). As analyzed by reverse transcriptase-polymerase chain reaction (RT-PCR), all the cell lines constitutively expressed COX-1, while COX-2 levels markedly varied among different cell lines. Induction of COX-2 by phorbol ester was observed in RPMI-8226 and HPC cells. In contrast, COX-2 was constitutively expressed in ARH-77 and IM-9 cells. Moreover, the high expression level of COX-2 protein in ARH-77 cells was verified by Western blotting. Intact cells of ARH-77 converted 14C-labeled arachidonic acid to prostaglandin E2, F2alpha, and D2, and this activity was dose-dependently inhibited by selective COX-2 inhibitors (SC-58125 and NS-398), a non-selective COX inhibitor (indomethacin), and relatively high concentrations of a selective COX-1 inhibitor (SC-560). These COX inhibitors also suppressed the proliferation of ARH-77 cells, but significant suppression was seen only at 100 microM, a much higher concentration than those sufficient for the COX inhibition. Moreover, proliferation of the myeloma cells lacking COX-2 was also suppressed by 100 microM of SC-58125. These results suggested that the anti-proliferative effect of the COX inhibitors is independent of the inhibition of COX-2.
Sigma-Aldrich
SC-58125, ≥98% (HPLC)