- [Determination of antidangdruff agent salicylic acid, zinc pyrithione, octopirox, climbazole and ketoconazole in shampoo by high performance liquid chromatography].
[Determination of antidangdruff agent salicylic acid, zinc pyrithione, octopirox, climbazole and ketoconazole in shampoo by high performance liquid chromatography].
Wei sheng yan jiu = Journal of hygiene research (2005-12-07)
Yan-Wei Yang, Ying Zhu, Xiao-Qing Su
PMID16329615
ABSTRACT
A high performance liquid chromatography method was established for determination of antidangdruff agent salicylic acid,zinc pyrithione, octopirox, climbazole and ketoconazole in shampoo on a C18 column using acetonitrile-metholaqueous solution (10 mmol/L KH2 PO4 and 5 mmol/L EDTANa2, pH is adjusted to 4.0 with H3 PO4) (50:10:40) as mobile phase at a flow rate of 1.0 ml/min, with the column temperature 25 degrees C and detection wave 230nm. The precision was less than 3.8% and recovery varied from 92.7% to 104.9%. The experimental results showed that the method was simple, precise and accurate.