Merck
  • Flufenacet herbicide treatment phenocopies the fiddlehead mutant in Arabidopsis thaliana.
Flufenacet herbicide treatment phenocopies the fiddlehead mutant in Arabidopsis thaliana.
Pest management science (2003-08-15)
Christa Lechelt-Kunze, Ruth C Meissner, Mark Drewes, Klaus Tietjen
ABSTRACT
In order to study the mode of action of herbicides we conducted a pilot study analysing phenotype and gene expression of flufenacet- and benfuresate-treated Arabidopsis thaliana (L) Heynhoe plants. Treatments with either herbicide caused phenocopies of the known Arabidopsis mutant fiddlehead, displaying fused organs and the typical fiddlehead-like inflorescence. Herbicide treatments of other plant species, including monocots, also gave rise to analogous organ fusions, indicating the presence of the target in a broad range of plants. Furthermore, many other herbicides with a proposed similar mode of action, eg chloroacetanilides, produced comparable fusion phenotypes in plants. The fiddlehead gene encodes a putative very-long-chain fatty acid elongase (VLCFAE), which corroborates earlier biochemical results pointing to the inhibition of VLCFA synthesis as mode of action of flufenacet. Gene expression profiles of herbicide-treated plants using the first 8247 gene Arabidopsis gene array of Affymetrix provided additional clues in support of inhibition of VLCFA synthesis. We discuss fiddlehead-like elongases as plant specific targets for flufenacet and many other herbicides.