- Stable conformations of tripeptides in aqueous solution studied by UV circular dichroism spectroscopy.
Stable conformations of tripeptides in aqueous solution studied by UV circular dichroism spectroscopy.
Determination of the precise solution structure of peptides is of utmost importance to the understanding of protein folding and peptide drugs. Herein, we have measured the UV circular dichroism (UVCD) spectra of tri-alanine dissolved in D(2)O, H(2)O, and glycerol. The results clearly show the coexistence of a polyproline II or 3(1)-helix and a somewhat disordered flat beta-strand conformation, in complete agreement with recent predictions from spectroscopic data (Eker et al. J. Am. Chem. Soc. 2002, 124, 14 330-14 341). A thermodynamic analysis revealed that enthalpic contributions of about 11 and 17 kJ/mol stabilize polyproline II in D(2)O and H(2)O, respectively, but at room temperature they are counterbalanced by entropic contributions, which clearly favor the more disordered beta-strand conformation. It is hypothesized that this delicate balance is the reason for the variety of structural propensities of amino acid residues in the absence of nonlocal interactions. The isotope effect yielding a higher occupation of polyproline II in H(2)O with respect to D(2)O strongly suggests that a hydrogen-bonding network involving the peptide and water molecules in the hydration shell plays a major role in stabilizing this conformation. The equilibrium between polyproline II and beta-strand is practically maintained in glycerol, which suggests that glycerol can substitute water as stabilizing solvent for the polyproline II conformation. We also measured the UVCD spectra of tri-valine and tri-lysine (both at acidic pD) in D(2)O and found them to adopt a flat beta-strand and left-handed turn structure, respectively, in accordance with recent analyses of vibrational spectroscopy data. Generally, the present study adds substantial evidence to the notion that the so-called random coil state of peptides is much more structured than generally assumed.