Merck
  • The antiviral activity of naturally occurring proteins and their peptide fragments after chemical modification.
The antiviral activity of naturally occurring proteins and their peptide fragments after chemical modification.
Antiviral research (2003-07-02)
Anna Oevermann, Monika Engels, Ursula Thomas, Antonio Pellegrini
ABSTRACT
Chemical modification of the proteins bovine serum albumin, alpha-lactalbumin, beta-lactoglobulin and chicken lysozyme by 3-hydroxyphthalic anhydride (3-HP) yielded compounds which exerted antiviral activity in vitro as compared with the native unmodified proteins. Of the three enveloped viruses tested, human herpes simplex virus type 1 (HSV-1), bovine parainfluenza virus type 3 and porcine respiratory corona virus, only HSV-1 proved sensitive to the 3-HP-proteins. All of the chemically modified proteins presented antiviral activity against HSV-1 when assayed before, during or after infection. However, to achieve HSV-1 inhibition, significantly higher concentrations of the modified proteins were required if present before infection as compared to during or after infection. Our results suggest that multiple mechanisms are involved in the inhibition of HSV-1 infection. Proteolytical digestion of albumin, alpha-lactalbumin, beta-lactoglobulin and lysozyme by trypsin, chymotrypsin and pepsin yielded several peptide fragments with antiherpetic activity. Chemical modification of these peptide fragments by 3-HP generated peptides with antiviral activity, however, this was almost always combined with a cytotoxic effect on the Vero cells. Overall, our results suggest that targeted chemical modification of some natural products might provide compounds effective against HSV-1 infection.
Sigma-Aldrich
3-Hydroxyphthalic anhydride, 98%