Merck
  • Efficacy of a combination febantel-praziquantel-pyrantel product, with or without vaccination with a commercial Giardia vaccine, for treatment of dogs with naturally occurring giardiasis.
Efficacy of a combination febantel-praziquantel-pyrantel product, with or without vaccination with a commercial Giardia vaccine, for treatment of dogs with naturally occurring giardiasis.
Journal of the American Veterinary Medical Association (2002-02-07)
Patricia A Payne, Robert K Ridley, Michael W Dryden, Christine Bathgate, George A Milliken, Patricia W Stewart
ABSTRACT
To determine efficacy of treatment with a combination febantel-praziquantel-pyrantel product, with or without vaccination with a commercial Giardia vaccine, in dogs with naturally occurring giardiasis. Prospective trial. 16 Beagles naturally infected with Giardia duodenalis. During phase 1, 6 dogs were treated with the parasiticide for 3 days (4 were also vaccinated). Four weeks later, all 6 dogs were treated with the parasiticide again for 5 days and were bathed and moved to clean cages after the last treatment (phase 2). Nine dogs were treated with the parasiticide for 3 (n = 4) or 5 (5) days and bathed and moved to clean cages after the last treatment (phase 3). Fecal samples were collected twice weekly for 24 days after treatment and tested for cysts with a quantitative zinc sulfate flotation technique and for Giardia antigen with an immunoassay. Dogs in phase 1 were all shedding cysts again by day 24. In phase 2, only 1 dog shed cysts after treatment, and shedding was transient (day 17). In phase 3, neither cysts nor antigen was detected in fecal samples from 2 of 4 dogs treated for 3 days and 4 of 5 dogs treated for 5 days. In 18 of 57 (31.6%) fecal samples, cysts were seen, but results of the immunoassay were negative. Results suggest that when a combination febantel-praziquantel-pyrantel product is used to treat dogs with giardiasis, bathing and changing the environment after treatment may be more important in preventing recurrence than duration of treatment.