Merck
  • Essential contribution of germline-encoded lysine residues in Jgamma1.2 segment to the recognition of nonpeptide antigens by human gammadelta T cells.
Essential contribution of germline-encoded lysine residues in Jgamma1.2 segment to the recognition of nonpeptide antigens by human gammadelta T cells.
Journal of immunology (Baltimore, Md. : 1950) (2001-12-12)
F Miyagawa, Y Tanaka, S Yamashita, B Mikami, K Danno, M Uehara, N Minato
ABSTRACT
Human gammadelta T cells display unique repertoires of Ag specificities largely imposed by selective usages of distinct Vgamma and Vdelta genes. Among them, Vgamma2/Vdelta2(+) T cells predominate in the circulation of healthy adults and respond to various microbial small molecular mass nonpeptide Ags. The present results indicate that the primary Vgamma2/Vdelta2(+) T cells stimulated with the distinct groups of nonpeptide Ags, including monoethyl pyrophosphate, isobutyl amine, and aminobisphosphonate, invariably exhibit Jgamma1.2 in the Vgamma2(+) TCR-gamma chains. Gene transfer studies revealed that most of the randomly cloned Vgamma2/Jgamma1.2(+) TCR-gamma genes bearing diverse Vgamma/Jgamma junctional sequences could confer the responsiveness to all these nonpeptide Ags, while none of the Vgamma2/Jgamma1.1(+) or Vgamma2/Jgamma1.3(+) TCR-gamma genes could do so. Furthermore, mutation of the lysine residues encoded by the Jgamma1.2 gene, which are unique in human Jgamma1.2 and absent in other human or mouse Jgamma segments, completely abrogated the responsiveness to all the nonpeptide Ags without affecting the response to anti-CD3 mAb. These results strongly suggested that the positively charged lysine residues in the TCR-gamma chain CDR3 region encoded by the germline Jgamma1.2 gene play a key role in the recognition of diverse small molecular mass nonpeptide Ags.
Sigma-Aldrich
Isobutylamine, 99%