Merck
  • Dopaminergic denervation enhances susceptibility to hydroxyl radicals in rat neostriatum.
Dopaminergic denervation enhances susceptibility to hydroxyl radicals in rat neostriatum.
Amino acids (2000-10-12)
R M Kostrzewa, J P Kostrzewa, R Brus
ABSTRACT
To determine if greater amounts of hydroxyl radical (*OH) are formed by dopamine (DA) denervation and treatment with L-dihydroxyphenylalanine (L-DOPA), the neostriatum was DA denervated (99% reduction in DA content) by 6-hydroxydopamine treatment (134microg icv, desipramine pretreatment) of neonatal rats. At 10 weeks the peripherally restricted dopa decarboxylase inhibitor carbidopa (12.5mg/kg i.p.) was administered 30min before vehicle, L-DOPA (60mg/kg i.p.), or the known generator of reactive oxygen species, 6-hydroxydopa (6-OHDOPA) (60mg/kg i.p.); and this was followed 30min later (and 15 min before termination) by the spin trap, salicylic acid (8micromoles icv). By means of a high performance liquid chromatographic method with electrochemical detection, we found a 4-fold increase in the non-enzymatically formed spin trap product, 2,3-dihydroxybenzoic acid (2,3-DHBA), with neither L-DOPA nor 6-OHDOPA having an effect on 2,3-DHBA content of the neostriatum. Basal content of 2,5-DHBA, the enzymatically formed spin trap product, was 4-fold higher vs. 2,3-DHBA in the neostriatum of untreated rats, while L-DOPA and 6-OHDOPA each reduced formation of 2,5-DHBA. We conclude that DA innervation normally suppresses *OH formation, and that the antiparkinsonian drug L-DOPA has no effect (2,3-DHBA) or slightly reduces (2,5-DHBA) *OH formation in the neostriatum, probably by virtue of its bathing the system of newly formed *OH.
Sigma-Aldrich
6-Hydroxy-DL-DOPA, ≥98% (HPLC), powder
USP
Levadopa Related Compound A, United States Pharmacopeia (USP) Reference Standard