Merck
  • Field evaluation of deltamethrin/S-bioallethrin/piperonyl butoxide and cyfluthrin against dengue vectors in Malaysia.
Field evaluation of deltamethrin/S-bioallethrin/piperonyl butoxide and cyfluthrin against dengue vectors in Malaysia.
Journal of vector ecology : journal of the Society for Vector Ecology (2000-08-05)
S Sulaiman, Z A Pawanchee, H F Othman, J Jamal, A Wahab, A R Sohadi, A Pandak
ABSTRACT
Deltacide (S-bioallethrin 0.71% w/v, deltamethrin 0.5% w/v, piperonyl butoxide 8.9% w/v excipients to 100% w/v) and Solfac UL 015 (cyfluthrin 1.5% w/v) were evaluated against the sentinel sugar-fed adults and 4th-instar larvae of Aedes aegypti at 17 storey high-rise apartments in Malaysia using ULV applications. The impact of both insecticides on field populations of Ae. aegypti and Ae. albopictus larvae was monitored weekly using bottle containers. Both Deltacide and Solfac UL 015 showed adulticidal and larvicidal effects. This was the first field trial using Deltacide against dengue vectors in Malaysia and showed its potential for use in dengue vector control programs.