Merck
  • [C677T and A1298C MTHFR polymorphisms in the etiology of neural tube defects in Spanish population].
[C677T and A1298C MTHFR polymorphisms in the etiology of neural tube defects in Spanish population].
Medicina clinica (2003-04-12)
José Ignacio Gutiérrez Revilla, Flora Pérez Hernández, M Teresa Calvo Martín, Manuel Tamparillas Salvador, Jesús Gracia Romero
ABSTRACT
The etiology of neural tube defects (NTDs) is multifactorial. The presence of mutated genotypes of C677T and A1298C polymorphisms, and their combined heterozygosity, have been considered risk factors for the occurrence and recurrence of NTDs in some populations. This case-control study included 159 healthy controls, 27 NTDs patients, 28 patients' mothers and 23 siblings. The polymorphism study was performed by PCR. For fragment digestion, we used the restriction enzymes Hinf I (C677T) and Mbo II (A1298C). There was no significant difference (p = 0.991) in C677T genotypes between controls (CC: 35%, CT: 50% and TT: 15%) and patients (37, 52 and 11%, respectively), patients' mothers (39, 50 and 11%, respectively) and siblings (35, 48 and 17%, respectively). The prevalence of A1298C genotypes in controls (AA: 49%, AC: 45% and CC: 6%) was similar (p = 0.917) to the prevalence in patients (41, 56 and 4%, respectively), patients' mothers (43, 50 and 7%, respectively) and siblings (52, 39 and 9%, respectively). The absence of differences in the two polymorphisms between these groups makes us conclude that there is no association with NTDs in the Spanish population.