Merck
  • Graphene oxide as a sulfur immobilizer in high performance lithium/sulfur cells.
Graphene oxide as a sulfur immobilizer in high performance lithium/sulfur cells.
Journal of the American Chemical Society (2011-10-25)
Liwen Ji, Mumin Rao, Haimei Zheng, Liang Zhang, Yuanchang Li, Wenhui Duan, Jinghua Guo, Elton J Cairns, Yuegang Zhang, Liwen Ji, Mumin Rao, Haimei Zheng, Liang Zhang, Yuanchang Li, Wenhui Duan, Jinghua Guo, Elton J Cairns, Yuegang Zhang
ABSTRACT
The loss of sulfur cathode material as a result of polysulfide dissolution causes significant capacity fading in rechargeable lithium/sulfur cells. Here, we use a chemical approach to immobilize sulfur and lithium polysulfides via the reactive functional groups on graphene oxide. This approach enabled us to obtain a uniform and thin (around tens of nanometers) sulfur coating on graphene oxide sheets by a simple chemical reaction-deposition strategy and a subsequent low-temperature thermal treatment process. Strong interaction between graphene oxide and sulfur or polysulfides enabled us to demonstrate lithium/sulfur cells with a high reversible capacity of 950-1400 mA h g(-1), and stable cycling for more than 50 deep cycles at 0.1C (1C = 1675 mA g(-1)).
Sigma-Aldrich
Graphene oxide, 4 mg/mL, dispersion in H2O, avg. no. of layers, 1
Sigma-Aldrich
Graphene oxide, organic solvent dispersible