Merck
  • Structure of a peptidoglycan amidase effector targeted to Gram-negative bacteria by the type VI secretion system.
Structure of a peptidoglycan amidase effector targeted to Gram-negative bacteria by the type VI secretion system.
Cell reports (2012-07-21)
Seemay Chou, Nhat Khai Bui, Alistair B Russell, Katrina W Lexa, Taylor E Gardiner, Michele LeRoux, Waldemar Vollmer, Joseph D Mougous
ABSTRACT
The target range of a bacterial secretion system can be defined by effector substrate specificity or by the efficacy of effector delivery. Here, we report the crystal structure of Tse1, a type VI secretion (T6S) bacteriolytic amidase effector from Pseudomonas aeruginosa. Consistent with its role as a toxin, Tse1 has a more accessible active site than related housekeeping enzymes. The activity of Tse1 against isolated peptidoglycan shows its capacity to act broadly against Gram-negative bacteria and even certain Gram-positive species. Studies with intact cells indicate that Gram-positive bacteria can remain vulnerable to Tse1 despite cell wall modifications. However, interbacterial competition studies demonstrate that Tse1-dependent lysis is restricted to Gram-negative targets. We propose that the previously observed specificity for T6S against Gram-negative bacteria is a consequence of high local effector concentration achieved by T6S-dependent targeting to its site of action rather than inherent effector substrate specificity.
Sigma-Aldrich
Amidase from Pseudomonas aeruginosa, recombinant, expressed in E. coli, buffered aqueous glycerol solution, hydroxamate transferase ≥200 units/mg protein (biuret)