Merck
  • Melatonin protects rat thymus against oxidative stress caused by exposure to microwaves and modulates proliferation/apoptosis of thymocytes.
Melatonin protects rat thymus against oxidative stress caused by exposure to microwaves and modulates proliferation/apoptosis of thymocytes.
General physiology and biophysics (2013-03-28)
Dusan Sokolovic, Branka Djordjevic, Gordana Kocic, Andrej Veljkovic, Milena Marinkovic, Jelena Basic, Tatjana Jevtovic-Stoimenov, Zoran Stanojkovic, Danka M Sokolovic, Voja Pavlovic, Boris Djindjic, Dejan Krstic
ABSTRACT
The aim of the study was to evaluate the effect of melatonin on oxidative stress, DNA fragmentation, apoptsis and proliferation in thymus tissue of rats exposed to microwaves. Wistar rats were divided in four groups: I - treated with saline; II - treated with melatonin; III - microwaves exposed; IV - microwaves exposed and melatonin treated. Melatonin (2 mg/kg i.p.) was administered daily. Animals were sacrificed after 20, 40 and 60 days. A significant increase in malondialdehyde and carbonyl group content, as well as decrease in catalase and increase in xanthine oxidase activity were registered under microwave exposure. Melatonin prevented the increase in malondialdehyde and carbonyl group content, and reversed the effect on catalase and xanthine oxidase activity. Both, alkaline and acid DNase activity were increased due to microwave exposure. Furthermore, microwaves caused increase in apoptosis rate (detected using Annexin V-FITC/PI kit) and reduced proliferative capacity of thymocytes (induced by ConA). However, melatonin caused decrease in alkaline and acid DNase activity, decrease in apoptotic rate and increase in proliferation rate of thymocytes. Melatonin exerts protective effects on rat thymocytes by modulating processes of apoptosis and proliferation, and causes decrease in DNA fragmentation and oxidative stress intensity under exposure to microwaves.
Sigma-Aldrich
Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade I, ammonium sulfate suspension, ≥0.4 units/mg protein
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade III, ammonium sulfate suspension, ≥0.8 units/mg protein
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade IV, ammonium sulfate suspension, ≥0.1 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Xanthine Oxidase microbial, lyophilized powder, ≥7 units/mg solid
Sigma-Aldrich
Xanthine Oxidase from bovine milk, lyophilized powder, 0.4-1.0 units/mg protein