Merck
  • GLP-1 secretion by microglial cells and decreased CNS expression in obesity.
GLP-1 secretion by microglial cells and decreased CNS expression in obesity.
Journal of neuroinflammation (2012-12-25)
Camilla Kappe, Linda M Tracy, Cesare Patrone, Kerstin Iverfeldt, Åke Sjöholm
ABSTRACT
Type 2 diabetes (T2D) is a strong risk factor for developing neurodegenerative pathologies. T2D patients have a deficiency in the intestinal incretin hormone GLP-1, which has been shown to exert neuroprotective and anti-inflammatory properties in the brain. Here we investigate potential sources of GLP-1 in the CNS and the effect of diabetic conditions on the proglucagon mRNA expression in the CNS. The obese mouse model ob/ob, characterized by its high levels of free fatty acids, and the microglia cell line BV-2 were used as models. mRNA expression and protein secretion were analyzed by qPCR, immunofluorescence and ELISA. We show evidence for microglia as a central source of GLP-1 secretion. Furthermore, we observed that expression and secretion are stimulated by cAMP and dependent on microglial activation state. We also show that insulin-resistant conditions reduce the central mRNA expression of proglucagon. The findings that microglial mRNA expression of proglucagon and GLP-1 protein expression are affected by high levels of free fatty acids and that both mRNA expression levels of proglucagon and secretion levels of GLP-1 are affected by inflammatory stimuli could be of pathogenic importance for the premature neurodegeneration and cognitive decline commonly seen in T2D patients, and they may also be harnessed to advantage in therapeutic efforts to prevent or treat such disorders.
Sigma-Aldrich
L-Arginase from bovine liver, Protein ≥70 % by biuret, powder
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Sigma-Aldrich
Chitinase from Streptomyces griseus, lyophilized powder (essentially salt free), ≥200 units/g solid
Sigma-Aldrich
Chitinase from Trichoderma viride, lyophilized powder, ≥600 units/g solid