Merck
  • Synthesis and biological evaluation of novel amides of polyunsaturated fatty acids with dopamine.
Synthesis and biological evaluation of novel amides of polyunsaturated fatty acids with dopamine.
Bioorganic & medicinal chemistry letters (2001-03-07)
V Bezuglov, M Bobrov, N Gretskaya, A Gonchar, G Zinchenko, D Melck, T Bisogno, V Di Marzo, D Kuklev, J C Rossi, J P Vidal, T Durand
ABSTRACT
New amides of different fatty acids from the C18, C20, and C22 series with dopamine were synthesized. Pharmacological characterization in binding assays with rat brain membrane preparations and in the 'tetrad' of cannabinoid behavioral tests showed that, for these compounds, cannabinoid-like activity was dependent on the fatty acid moiety. Our data demonstrate that polyenoic fatty acid amides with dopamine comprise a new family of synthetic cannabimimetics.
Sigma-Aldrich
Arachidonoyl dopamine, ≥98% (HPLC), ethanol solution