Merck
  • Duration of ergovaline exposure influences serotonin-mediated vasoactivity of bovine mesenteric vasculature.
Duration of ergovaline exposure influences serotonin-mediated vasoactivity of bovine mesenteric vasculature.
Journal of animal science (2023-04-03)
Ronald J Trotta, David L Harmon, Huihua Ji, James L Klotz
ABSTRACT
Ergovaline (ERV), produced in toxic endophyte-infected tall fescue, causes potent vasoconstriction of bovine peripheral and visceral vasculature. Ergovaline acts as both an agonist and an antagonist in bovine gut blood vessels through serotonin (5-HT) receptors and it appears that the type of action could be influenced by the extent of ERV exposure. Because it was unclear how the duration of ERV exposure influences 5-HT-mediated vasoactivity, experiments were designed to evaluate how simultaneous or prior ERV exposure influenced 5-HT-mediated vasoactivity of mesenteric artery (MA) and vein (MV) segments from Holstein steers (N = 10). Vessels were incubated in Krebs-Henseleit buffer containing 0, 0.01, or 0.1 μM ERV for 24 h prior to the 5-HT dose-response or exposed to fixed concentrations of 0, 0.01, or 0.1 μM ERV simultaneously during the 5-HT dose-response. Vessels were suspended in chambers of a multimyograph containing Krebs-Henseleit buffer and equilibrated to 1 g tension for 90 min. Vessels were exposed to increasing concentrations of 5-HT (5 × 10-8 M to 1 × 10-4 M) every 15 min and contractile responses were normalized as a percentage of the maximum contractile response induced by 120 mM KCl reference addition. Two-way analysis of variance was used to separately analyze data for each vessel type and duration of exposure using the MIXED procedure of SAS. When 5-HT concentration increased from 5 × 10-8 to 1 × 10-6 M, simultaneous addition of 0.1 μM ERV increased (P < 0.01) the contractile response of MV compared with additions of 0 and 0.01 μM ERV. At 1 × 10-4 M 5-HT, the simultaneous presence of 0.01 and 0.1 μM ERV decreased (P < 0.01) the contractile response of both MA and MV compared with 0 μM ERV addition. As 5-HT concentrations increased, the contractile response increased (P < 0.01) in both MA and MV with no previous ERV exposure, but decreased in MA and MV with 24 h prior exposure to 0.01 and 0.1 μM ERV. These data demonstrate that the duration of ERV exposure influences 5-HT-mediated vasoconstriction and likely vasorelaxation in bovine mesenteric vasculature. If ERV and 5-HT exposure occur simultaneously, ERV can act as a partial agonist of 5-HT-mediated vasoconstriction. If 5-HT exposure occurs after blood vessels have had prior ERV exposure, it appears that 5-HT may induce vasorelaxation of blood vessels. More research is needed to identify cellular and molecular mechanisms involved with 5-HT-mediated vasoactivity.
Sigma-Aldrich
Serotonin hydrochloride, powder
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-1250242-100MG100 mg
Ships in 14 days - from Aldrich Partner.
€406.40
EN300-1250242-250MG250 mg
Ships in 14 days - from Aldrich Partner.
€562.40
EN300-1250242-500MG500 mg
Ships in 14 days - from Aldrich Partner.
€857.50
EN300-1250242-1G1 g
Ships in 14 days - from Aldrich Partner.
€1,087.60
EN300-1250242-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
€2,088.60
EN300-1250242-5G5 g
Ships in 14 days - from Aldrich Partner.
€3,068.80
EN300-1250242-10G10 g
Ships in 14 days - from Aldrich Partner.
€4,530.00