Merck
  • Elucidation of E3 ubiquitin ligase specificity through proteome-wide internal degron mapping.
Elucidation of E3 ubiquitin ligase specificity through proteome-wide internal degron mapping.
Molecular cell (2023-09-23)
Zhiqian Zhang, Brandon Sie, Aiquan Chang, Yumei Leng, Christopher Nardone, Richard T Timms, Stephen J Elledge
ABSTRACT
The ubiquitin-proteasome system plays a critical role in biology by regulating protein degradation. Despite their importance, precise recognition specificity is known for a few of the 600 E3s. Here, we establish a two-pronged strategy for identifying and mapping critical residues of internal degrons on a proteome-scale in HEK-293T cells. We employ global protein stability profiling combined with machine learning to identify 15,800 peptides likely to contain sequence-dependent degrons. We combine this with scanning mutagenesis to define critical residues for over 5,000 predicted degrons. Focusing on Cullin-RING ligase degrons, we generated mutational fingerprints for 219 degrons and developed DegronID, a computational algorithm enabling the clustering of degron peptides with similar motifs. CRISPR analysis enabled the discovery of E3-degron pairs, of which we uncovered 16 pairs that revealed extensive degron variability and structural determinants. We provide the visualization of these data on the public DegronID data browser as a resource for future exploration.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody