Merck
  • Differential expression of interleukin-35 receptor distinguishes different subsets of granulocyte-macrophage-colony-stimulating factor-producing T helper cells in a mouse endometriosis model.
Differential expression of interleukin-35 receptor distinguishes different subsets of granulocyte-macrophage-colony-stimulating factor-producing T helper cells in a mouse endometriosis model.
Molecular immunology (2023-11-09)
Fengqin Lin, Hongbo Yu, Li Zhang, Jing Zhou, Yuan Cao, Songli Wu, Junjie Wang
ABSTRACT
The immune system contributes to the pathophysiology of endometriosis. The role of ThGM cells, which produce granulocyte macrophage-colony-stimulating factor (GM-CSF), in the pathogenesis of endometriosis remains unknown. To analyze the features of ThGM cells in endometriosis, a mouse endometriosis model was established. ThGM cells in the spleen, peritoneal fluid (PF), and endometriotic lesions (EL) were measured by flow cytometry, based on the expression of surface markers and intracellular proteins. Live ThGM cells were sorted according to chemokine receptor expression profiles and their effects on other CD4+ T cell subsets were determined by co-culture assays. An adoptive transfer assay was performed to characterize the effect of ThGM cells on endometriosis. We found that ThGM cells were present in endometriotic PF and EL. Live EL ThGM cells were enriched in CD4+CXCR3-CCR8-CCR4+CCR10+ T cells. EL ThGM cells differentially express interleukin-35 receptor (IL-35R), consisting of an IL-35R+ subset and an IL-35R- subset. The IL-35R+ subset expressed less GM-CSF, interleukin-2 (IL-2), and tumor necrosis factor-alpha (TNF-α) and proliferated slower than the IL-35R- subset. Meanwhile, the IL-35R+ subset was weaker than the IL-35R- subset in promoting the functions of Th1 and Th17 cells. ThGM cell transfer did not influence EL development but significantly alleviated pro-inflammatory cytokines in PF and ELs. Interleukin-35 (IL-35), the ligand of IL-35R, suppressed ThGM cell function and proliferation in an IL-35R-dependent manner. In summary, ThGM cells in the PF and ELs might exacerbate endometriotic inflammation. IL-35 might suppress the function of ThGM cells via IL-35R.
Sigma-Aldrich
IL-35 (mouse): FC (human), recombinant, expressed in CHO cells