Merck
  • Quantifying Intracellular Nanoparticle Distributions with Three-Dimensional Super-Resolution Microscopy.
Quantifying Intracellular Nanoparticle Distributions with Three-Dimensional Super-Resolution Microscopy.
ACS nano (2023-04-19)
Vinit Sheth, Xuxin Chen, Evan M Mettenbrink, Wen Yang, Meredith A Jones, Ons M'Saad, Abigail G Thomas, Rylee S Newport, Emmy Francek, Lin Wang, Alex N Frickenstein, Nathan D Donahue, Alyssa Holden, Nathan F Mjema, Dixy E Green, Paul L DeAngelis, Joerg Bewersdorf, Stefan Wilhelm
ABSTRACT
Super-resolution microscopy can transform our understanding of nanoparticle-cell interactions. Here, we established a super-resolution imaging technology to visualize nanoparticle distributions inside mammalian cells. The cells were exposed to metallic nanoparticles and then embedded within different swellable hydrogels to enable quantitative three-dimensional (3D) imaging approaching electron-microscopy-like resolution using a standard light microscope. By exploiting the nanoparticles' light scattering properties, we demonstrated quantitative label-free imaging of intracellular nanoparticles with ultrastructural context. We confirmed the compatibility of two expansion microscopy protocols, protein retention and pan-expansion microscopy, with nanoparticle uptake studies. We validated relative differences between nanoparticle cellular accumulation for various surface modifications using mass spectrometry and determined the intracellular nanoparticle spatial distribution in 3D for entire single cells. This super-resolution imaging platform technology may be broadly used to understand the nanoparticle intracellular fate in fundamental and applied studies to potentially inform the engineering of safer and more effective nanomedicines.
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)