Merck
  • ISL1/SHH/CXCL12 signaling regulates myogenic cell migration during mouse tongue development.
ISL1/SHH/CXCL12 signaling regulates myogenic cell migration during mouse tongue development.
Development (Cambridge, England) (2022-10-06)
Wei Zhang, Jiaojiao Yu, Guoquan Fu, Jianying Li, Huarong Huang, Jing Liu, Dongliang Yu, Mengsheng Qiu, Feixue Li
ABSTRACT
Migration of myoblasts derived from the occipital somites is essential for tongue morphogenesis. However, the molecular mechanisms of myoblast migration remain elusive. In this study, we report that deletion of Isl1 in the mouse mandibular epithelium leads to aglossia due to myoblast migration defects. Isl1 regulates the expression pattern of chemokine ligand 12 (Cxcl12) in the first branchial arch through the Shh/Wnt5a cascade. Cxcl12+ mesenchymal cells in Isl1ShhCre embryos were unable to migrate to the distal region, but instead clustered in a relatively small proximal domain of the mandible. CXCL12 serves as a bidirectional cue for myoblasts expressing its receptor CXCR4 in a concentration-dependent manner, attracting Cxcr4+ myoblast invasion at low concentrations but repelling at high concentrations. The accumulation of Cxcl12+ mesenchymal cells resulted in high local concentrations of CXCL12, which prevented Cxcr4+ myoblast invasion. Furthermore, transgenic activation of Ihh alleviated defects in tongue development and rescued myoblast migration, confirming the functional involvement of Hedgehog signaling in tongue development. In summary, this study provides the first line of genetic evidence that the ISL1/SHH/CXCL12 axis regulates myoblast migration during tongue development.