Merck
  • Wfs1E864K knock-in mice illuminate the fundamental role of Wfs1 in endocochlear potential production.
Wfs1E864K knock-in mice illuminate the fundamental role of Wfs1 in endocochlear potential production.
Cell death & disease (2023-06-30)
Elodie M Richard, Emilie Brun, Julia Korchagina, Lucie Crouzier, Corentin Affortit, Stacy Alves, Chantal Cazevieille, Anne-Laure Mausset-Bonnefont, Marc Lenoir, Jean-Luc Puel, Tangui Maurice, Marc Thiry, Jing Wang, Benjamin Delprat
ABSTRACT
Wolfram syndrome (WS) is a rare neurodegenerative disorder encompassing diabetes mellitus, diabetes insipidus, optic atrophy, hearing loss (HL) as well as neurological disorders. None of the animal models of the pathology are presenting with an early onset HL, impeding the understanding of the role of Wolframin (WFS1), the protein responsible for WS, in the auditory pathway. We generated a knock-in mouse, the Wfs1E864K line, presenting a human mutation leading to severe deafness in affected individuals. The homozygous mice showed a profound post-natal HL and vestibular syndrome, a collapse of the endocochlear potential (EP) and a devastating alteration of the stria vascularis and neurosensory epithelium. The mutant protein prevented the localization to the cell surface of the Na+/K+ATPase β1 subunit, a key protein for the maintenance of the EP. Overall, our data support a key role of WFS1 in the maintenance of the EP and the stria vascularis, via its binding partner, the Na+/K+ATPase β1 subunit.