Merck
  • Evaluation of Dilution Susceptibility Testing Methods for Aztreonam in Combination with Avibactam against Enterobacterales.
Evaluation of Dilution Susceptibility Testing Methods for Aztreonam in Combination with Avibactam against Enterobacterales.
Microbiology spectrum (2022-11-08)
C J Kelley, A Kennedy-Mendez, O N Walser, M T Thwaites, F F Arhin, C M Pillar, D A Hufnagel
ABSTRACT
As multidrug and pan-resistance among Enterobacterales continue to increase, there is an urgent need for more therapeutic options to treat these infections. New β-lactam and β-lactam inhibitor (BLI) combinations have a broad spectrum of activity, but those currently approved do not provide coverage against isolates harboring metallo-β-lactamases (MBL). Aztreonam (ATM) and avibactam (AVI) in combination (ATM/AVI; AVI at 4 μg/mL fixed concentration) provides a similarly broad range of activity while maintaining activity against MBL-producing isolates. The in vitro susceptibility testing of ATM/AVI by standard methods was evaluated during development. This study investigated the impact of nonstandard testing conditions on the activity of ATM/AVI as observed during broth microdilution testing as well as the equivalency between agar dilution and broth microdilution MIC values when testing a diverse panel of Enterobacterales (N = 201). Nonstandard test conditions evaluated included inoculum density, atmosphere of incubation, media pH, varied medium cation concentrations, incubation time, varied serum concentrations, testing in pooled urine instead of media, addition of blood to the media, and the presence of surfactant. Generally, apart from low pH and high inoculum density, nonstandard testing parameters did not affect ATM/AVI broth microdilution MIC values. Correlation of MIC values obtained by agar dilution and broth microdilution resulted in an essential agreement of 97.0% for all tested Enterobacterales. Variation of standard testing conditions had little impact on broth microdilution MIC values for ATM/AVI. The correlation between broth microdilution and agar dilution MICs suggests both methods are reliable for determination of ATM/AVI MIC values. IMPORTANCE Increasing antibiotic resistance and emergence of pan-resistant isolates threaten the ability to control infections and to provide many other medical interventions such as surgery and chemotherapy, among others. New therapies are required to control emerging resistance mechanisms, including the increase in metallo-β-lactamases. Some new antibiotic combinations provide coverage against highly resistant isolates but are unable to target organisms that produce metallo-β-lactamases. Aztreonam in combination with avibactam provides a broad spectrum of activity against highly resistant isolates that also targets metallo-β-lactamase-producing organisms. An important part of drug development is the ability for clinical labs to determine the susceptibility of isolates to the antimicrobial. This manuscript investigates the in vitro susceptibility testing of aztreonam/avibactam with nonstandard testing conditions and a correlation study between broth microdilution and agar dilution against clinical isolates encoding a variety of resistance mechanisms. Overall, aztreonam/avibactam was generally unaffected by changes in testing conditions and showed strong agar/broth correlation.