Merck
  • ATF1 promotes the malignancy of lung adenocarcinoma cells by transcriptionally regulating ZNF143 expression.
ATF1 promotes the malignancy of lung adenocarcinoma cells by transcriptionally regulating ZNF143 expression.
Acta biochimica et biophysica Sinica (2023-05-09)
Jinhong Mei, Yu Liu, Yiyun Sheng, Ying Liu, Limin Chen, Hailong Wang, Minzhang Cheng, Zhenyu Zhai, Linlin Xu
ABSTRACT
The clinical oncogenic functions and mechanisms of activating transcription factor 1 (ATF1) in the progression of lung adenocarcinoma have not been completely elucidated. In this study, by employing human lung adenocarcinoma tissues and cells, we detect the correlation of ATF1 expression with the clinicopathological features and prognosis of patients with lung adenocarcinoma and find that ATF1 promotes lung adenocarcinoma cell proliferation and migration by transcriptionally enhancing zinc finger protein 143 (ZNF143) expression. ATF1 and ZNF143 are strongly expressed in lung adenocarcinoma tissues compared with those in the adjacent normal tissues, and high ATF1 and ZNF143 expressions are related to poor disease-free survival of lung adenocarcinoma patients. ATF1 overexpression results in increased proliferation and migration of lung adenocarcinoma cells, whereas knockdown of ATF1 inhibits cell proliferation and migration. Furthermore, ATF1 transcriptionally regulates the expression of ZNF143, and ATF1 and ZNF143 expressions are positively correlated in lung adenocarcinoma tissues. ZNF143 knockdown blocks lung adenocarcinoma cell migration, which is mediated by ATF1 upregulation. Hence, this study provides a potential therapeutic candidate for the treatment of lung adenocarcinoma.
Sigma-Aldrich
Anti-ZNF143 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution