Merck
  • DHA Induces Cell Death through the Production of ROS and the Upregulation of CHOP in Fibroblast-like Synovial Cells from Human Rheumatoid Arthritis Patients.
DHA Induces Cell Death through the Production of ROS and the Upregulation of CHOP in Fibroblast-like Synovial Cells from Human Rheumatoid Arthritis Patients.
International journal of molecular sciences (2023-01-22)
Mini Jeong, Jong-Il Shin, Jaewook Cho, Yong-Joon Jeon, Jin-Hyun Kim, Jeehee Youn, Kyungho Lee
ABSTRACT
Rheumatoid arthritis (RA) is an inflammatory disease marked by a massive proliferation of synovial cells in the joints. In this study, we investigated the pro-apoptotic effects of docosahexaenoic acid (DHA) in human fibroblast-like synovial cells from RA patients (RA-FLS). An in vitro study using MH7A cells showed that DHA treatment induced caspase-8-dependent apoptosis in a dose-dependent manner and reduced the TNF-α-mediated induction of MMP-9 and IL-1β. DHA also induced the phosphorylation of eIF2α, the expression of the ER stress markers ATF4 and C/EBP homologous protein (CHOP), and death receptor 5 (DR5). The knockdown of CHOP or DR5 increased cell viability and reduced apoptosis in DHA-treated cells. Furthermore, the knockdown of CHOP reduced DHA-mediated DR5 expression, while the overexpression of CHOP increased DR5 expression. We also found that DHA treatment induced the accumulation of reactive oxygen species (ROS), and pretreatment with the anti-oxidant Tiron effectively abrogated not only the expression of CHOP and DR5, but also DHA-induced apoptosis. Under this condition, cell viability was increased, while PARP-1 cleavage and caspase-8 activation were reduced. All the findings were reproduced in human primary synovial cells obtained from RA patients. These results suggest that the DHA-mediated induction of ROS and CHOP induced apoptosis through the upregulation of DR5 in RA-FLSs, and that CHOP could be used as a therapy for RA.