- A highly luminescent spiro-anthracenone-based organic light-emitting diode exhibiting thermally activated delayed fluorescence.
A highly luminescent spiro-anthracenone-based organic light-emitting diode exhibiting thermally activated delayed fluorescence.
Chemical communications (Cambridge, England) (2013-09-28)
Keiro Nasu, Tetsuya Nakagawa, Hiroko Nomura, Chi-Jen Lin, Chien-Hong Cheng, Mei-Rurng Tseng, Takuma Yasuda, Chihaya Adachi
PMID24071893
ABSTRACT
Efficient thermally activated delayed fluorescence was observed from a spiro-anthracenone derivative (ACRSA). An organic light-emitting diode containing ACRSA exhibited blue-greenish electroluminescence with a maximum external quantum efficiency of 16.5%.