Merck
  • Pericarditis mediated by respiratory syncytial virus in a hematopoietic stem cell transplant patient.
Pericarditis mediated by respiratory syncytial virus in a hematopoietic stem cell transplant patient.
Transplant infectious disease : an official journal of the Transplantation Society (2013-06-04)
M P Rubach, E N Pavlisko, J R Perfect
ABSTRACT
We describe a case of pericarditis and large pericardial effusion in a 63-year-old African-American man undergoing autologous hematopoietic stem cell transplant for multiple myeloma. Pericardial tissue biopsy demonstrated fibrinous pericarditis, and immunohistochemistry stains were positive for respiratory syncytial virus. The patient improved with oral ribavirin and intravenous immune globulin infusions.