Merck
  • Genotypic variation and slime production among blood and catheter isolates of Candida parapsilosis.
Genotypic variation and slime production among blood and catheter isolates of Candida parapsilosis.
Journal of clinical microbiology (1994-02-01)
M L Branchini, M A Pfaller, J Rhine-Chalberg, T Frempong, H D Isenberg
ABSTRACT
Candida parapsilosis is an important nosocomial pathogen that can proliferate in high concentrations of glucose and form biofilms on prosthetic materials. We investigated the genotypic diversity and slime production among 31 isolates of C. parapsilosis from individual patients with bloodstream or catheter infections. DNA subtyping was performed by using electrophoretic karyotyping plus restriction endonuclease analysis with BssHII followed by pulsed-field gel electrophoresis. Slime production was evaluated by growing organisms in Sabouraud broth with 8% glucose and examining the walls of the tubes for the presence of an adherent slime layer. Overall there were 14 DNA subtypes among the 31 isolates. Eighty percent of the isolates produced slime; 67% of the isolates were moderately to strongly positive, 13% were weakly positive, and 20% were not slime producers. The ability of isolates of a given DNA type to produce slime under these conditions was variable. The results of these studies indicate moderate genotypic variation among clinical isolates of C. parapsilosis. The propensity of these isolates to form slime in glucose-containing solutions suggests that this phenotypic characteristic may contribute to the ability of C. parapsilosis to adhere to plastic catheters and cause infections.