Merck
Intraretinal pigmented cells in retinal degenerative disease.
The British journal of ophthalmology (2022-03-19)
Marina Yasvoina, Qian Yang, Sasha M Woods, Tjebo Heeren, Grant M Comer, Catherine A Egan, Marcus Fruttiger
ABSTRACT
Invasion of pigmented cells into the retina occurs in retinal degenerative diseases, such as macular telangiectasia type 2 (MacTel) and retinitis pigmentosa (RP). These intraretinal pigmented cells may be derived from the retinal pigment epithelium (RPE), but differences and similarities between intraretinal pigmented cells and RPE have so far not been well characterised.Clinicopathologic case report. Here, we compared intraretinal pigment cells with RPE cells by immunohistochemistry. Immunohistological stains for classic RPE markers (RPE65, CRALBP and KRT18) and blood vessel markers (lectin and collagen 4) were done on sections from postmortem eye tissue from two MacTel donors, an RP donor and a control donor. Presence of specific immunohistochemistry markers on intraretinal pigmented and RPE cells. We found that intraretinal pigmented cells did not express RPE65 and CRALBP, with a small subset expressing them weakly. However, they all expressed KRT18, which was also present in normal RPE cells. Interestingly, we also found clusters of KRT18-positive cells in the retina that were not pigmented. Our findings suggest that RPE cells invading the retina dedifferentiate (losing classic RPE markers) and can be pigmented or unpigmented. Therefore, the number of RPE cells invading the retina in retinal degenerative disease may be underappreciated by funduscopy.
Sigma-Aldrich
Monoclonal Anti-Vimentin−Cy3 antibody produced in mouse, clone V9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture