Merck
  • Coriander (Coriandrum sativum L.) essential oil and oil-loaded nano-formulations as an anti-aging potentiality via TGFβ/SMAD pathway.
Coriander (Coriandrum sativum L.) essential oil and oil-loaded nano-formulations as an anti-aging potentiality via TGFβ/SMAD pathway.
Scientific reports (2022-04-23)
Mohamed A Salem, Eman G Manaa, Nada Osama, Nora M Aborehab, Mai F Ragab, Yusuf A Haggag, Magda T Ibrahim, Dalia I Hamdan
ABSTRACT
Aging has become a concern for many people, especially women. Given that high-quality anti-aging products are of high cost; it has imperative to search for other economical sources. Essential oils are frequently used in cosmetics products due to a wide range of biological activities as well as their pleasant odor. The current study aimed to investigate the biochemical effect of the cosmetic potential of selected Apiaceous essential oils, traditionally used for skincare, by evaluating their anti-wrinkle activity. It is worth noting that, coriander essential oil showed the highest collagenase, elastase, tyrosinase, and hyaluronidase inhibitory activities compared to other Apiaceous oils (fennel, anise, and cumin). GC-MS proved that coriander essential oil showed a very high level of oxygenated monoterpenes, with linalool (81.29%) as the most abundant constituent. Intriguingly, coriander oil cream and Coriander Essential Oil-loaded Lipid Nanoparticles (CEOLNs) formulations attenuated in vivo UV-induced skin photoaging that was manifested by significantly decreased MDA, COX-2, PGE-2, MMP-1, JNK, and AP-1 levels. Moreover, these pharmaceutical dosage forms significantly increased skin collagen content compared to UV-injured group. Also, coriander essential oil significantly increased TGFβ, TGFβII, and SMAD3 protein expression levels compared to UV-injured group. In conclusion, the pharmaceutical dosage forms of coriander oil possess anti-wrinkle activities that could have an auspicious role in amending extrinsic aging.
Sigma-Aldrich
Anti-AP-1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution