Merck
  • Novel Human Full-Thickness Three-Dimensional Nonkeratinized Mucous Membrane Model for Pharmacological Studies in Wound Healing.
Novel Human Full-Thickness Three-Dimensional Nonkeratinized Mucous Membrane Model for Pharmacological Studies in Wound Healing.
Skin pharmacology and physiology (2019-07-10)
Laurenz Schmitt, Yvonne Marquardt, Ruth Heise, Verena von Felbert, Philipp M Amann, Laura Huth, Timm Steiner, Frank Hölzle, Sebastian Huth, Jens Malte Baron
ABSTRACT
Efforts are increasingly aiming to develop in vitro models that can provide effective alternatives to in vivo experiments. The main aim of this study was the establishment of an in vitro model of the nonkeratinized mucous membrane that can be used as a standardized tool to evaluate biological and therapeutic effects of pharmaceuticals for mucosal wound healing. We established a full-thickness in vitro model of the nonkeratinized mucous membrane. While histological examination was performed to assess morphological characteristics, we utilized gene expression profiling using microarray and qRT-PCR analyses to identify molecular effects of treatment with a dexpanthenol-containing ointment after laser wounding. Performing histological and immunofluorescence analyses we proved that our model mimics the two distinctive layers of the mucous membrane - the stratified squamous epithelium and the lamina propria. We used this model to investigate molecular effects of a dexpanthenol-containing ointment that is commonly used for the wound treatment of mucous membranes. For that purpose, our model exhibits a unique feature in that dexpanthenol and proliferation-enhancing additives that may interfere with our studies are not required for the maintenance of the model culture. After setting standardized lesions with a nonsequential fractional ultrapulsed CO2 laser, topical treatment with the dexpanthenol-containing ointment enhanced wound closure in the model compared to placebo and untreated controls. Furthermore, microarray analysis revealed that the treatment of the laser-wounded model with the dexpanthenol-containing ointment evoked an upregulated expression of various genes related to accelerated wound healing. Overall, we verified that this novel mucous membrane model can be utilized in future to monitor ex vivo effects of various topical therapies on mucosa morphology, physiology, and gene expression. Our findings confirm the potential of the model as an in vitro tool for the replacement of pharmacological in vivo studies regarding mucosal wound healing.