Merck
  • Pharmacological characterization of BR-A-657, a highly potent nonpeptide angiotensin II receptor antagonist.
Pharmacological characterization of BR-A-657, a highly potent nonpeptide angiotensin II receptor antagonist.
Biological & pharmaceutical bulletin (2013-07-03)
Yong Ha Chi, Joo Han Lee, Je Hak Kim, Hyun Kwang Tan, Sang Lin Kim, Jae Yeol Lee, Hong-Kun Rim, Soo Heui Paik, Kyung-Tae Lee
ABSTRACT
The pharmacological profile of BR-A-657, 2-n-butyl-5-dimethylamino-thiocarbonyl-methyl-6-methyl-3-{[2-(1H-tetrazole-5-yl)biphenyl-4-yl]methyl}-pyrimidin-4(3H)-one, a new nonpeptide AT1-selective angiotensin receptor antagonist, has been investigated in a variety of in vitro and in vivo experimental models. In the present study, BR-A-657 displaced [(125)I][Sar(1)-Ile(8)]angiotensin II (Ang II) from its specific binding sites to AT1 subtype receptors in membrane fractions of HEK-293 cells with an IC50 of 0.16 nM. In a functional assay using isolated rabbit thoracic aorta, BR-A-657 inhibited the contractile response to Ang II (pD'2: 9.15) with a significant reduction in the maximum. In conscious rats, BR-A-657 (0.01, 0.1, 1 mg/kg; intravenously (i.v.)) dose-dependently antagonized Ang II-induced pressor responses. In addition, BR-A-657 dose-dependently decreased mean arterial pressure in furosemide-treated rats and renal hypertensive rats. Moreover, BR-A-657 given orally at 1 and 3 mg/kg reduced blood pressure in conscious renal hypertensive rats. Taken together, these findings indicate that BR-A-657 is a potent and specific antagonist of Ang II at the AT1 receptor subtype, and reveal the molecular basis responsible for the marked lowering of blood pressure in conscious rats.
Sigma-Aldrich
Fimasartan, ≥98% (HPLC)
Pricing and availability is not currently available.